Merck
  • A multi-residue method for the determination of seven polypeptide drug residues in chicken muscle tissues by LC-MS/MS.
A multi-residue method for the determination of seven polypeptide drug residues in chicken muscle tissues by LC-MS/MS.
Analytical and bioanalytical chemistry (2015-04-03)
Joe O Boison, Stephen Lee, Johanna Matus
ABSTRACT
A new multi-residue method for the determination of seven polypeptides, namely, polymixin B1, polymixin B2, polymixin E1 (colistin A), polymixin E2 (colistin B), enduracidin A (enramycin A), enduracidin B (enramycin B), and bacitracin A, in food of animal origin was developed and validated for chicken muscle tissue. Chicken muscle tissue was extracted with acidified methanol (1 % TFA). After homogenization, shaking, and centrifugation, the acidified methanol extract was decanted. A second extraction was performed with methanol (1 % TFA) and formic acid (1 %) 25:75, v/v. The pooled extract was cleaned up and concentrated on a solid-phase extraction cartridge. The retained analytes were eluted with methanol/acetonitrile. The extract was evaporated to dryness, reconstituted in mobile phase, filtered, and quantified by LC-MS/MS under ESI conditions. The method has a LOQ of 50.0 μg/kg for polymixin E2 (colistin B), 39.0 μg/kg for polymixin E1 (colistin A), 74.0 μg/kg for polymixin B1, 71.0 μg/kg for polymixin B2, 66.0 μg/kg for enduracidin A, 50.0 μg/kg for enduracidin B, and 30.0 μg/kg for bacitracin A in chicken muscle tissues. This is the first sensitive, suitable, multi-residue method reported for the seven polypeptide drug residues in chicken muscle tissue.
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis