Merck
  • Tissue engineering of nanosilver-embedded peripheral nerve scaffold to repair nerve defects under contamination conditions.
Tissue engineering of nanosilver-embedded peripheral nerve scaffold to repair nerve defects under contamination conditions.
The International journal of artificial organs (2015-10-21)
Tan Ding, Jun-Bin Yin, Hu-Ping Hao, Chao Zhu, Ting Zhang, Ya-Cheng Lu, Li-Ying Wang, Zhe Wang, Yun-Qing Li
ABSTRACT
We employed a nanosilver-collagen scaffold and tested its effects on inhibiting bacteria and facilitating nerve regeneration. Based on our previous research, we prepared bionic scaffolds with different concentrations of nanosilver and examined their internal structures by scanning electron microscopy and energy dispersive spectroscopy. We implanted these scaffolds or autologous nerve grafts into rats to repair a 10-mm injury of the sciatic nerve. The 2 mg/ml group showed a >10 mm bacterial inhibition zone in all 3 types of bacterial culture dishes. At day 60 postsurgery, the 2 mg/ml group also showed the highest amplitude of evoked potential (AMP) and nerve conduction velocity (NCV). The regenerating nerves in the 2 mg/ml group were denser and more mature, and with thicker and well-arrayed myelin sheath. These results demonstrate that nanosilver scaffolds (2 mg/ml group) were effective in inhibiting bacteria both in vitro and in vivo, and reduced the contamination-caused immune responses, which in turn promoted nerve regeneration and functional recovery.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase