Merck
  • Purification and characterization of four antibacterial peptides from protamex hydrolysate of Atlantic mackerel (Scomber scombrus) by-products.
Purification and characterization of four antibacterial peptides from protamex hydrolysate of Atlantic mackerel (Scomber scombrus) by-products.
Biochemical and biophysical research communications (2015-05-03)
Nadia Ennaas, Riadh Hammami, Lucie Beaulieu, Ismail Fliss
ABSTRACT
Proteins from fish by-product sources are valuable source of bioactive peptides and show promise as functional foods ingredients. The objective of the present study was to isolate and characterize antibacterial peptides from protamex hydrolysates of Atlantic mackerel (Scomber scombrus) by-products. Four sequences SIFIQRFTT (P4), RKSGDPLGR (P8.1), AKPGDGAGSGPR (P8.2) and GLPGPLGPAGPK (P11) were identified in peptide fractions separated using RP-HPLC. At 200 μg mL(-1), while peptides P8.1, P8.2 and P11 exhibited partial inhibition, P4 totally inhibited tested Gram-positive (Listeria innocua) and Gram-negative (Escherichia coli) bacterial strains. These results suggest that the protein hydrolysate derived from mackerel by-products could be used as an antimicrobial ingredient in both functional food and nutraceutical applications.
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)