Merck
  • Reducing pathogens in combined sewer overflows using ozonation or UV irradiation.
Reducing pathogens in combined sewer overflows using ozonation or UV irradiation.
International journal of hygiene and environmental health (2015-10-04)
Katharina Tondera, Kassandra Klaer, Jens Gebhardt, Jost Wingender, Christoph Koch, Marina Horstkott, Martin Strathmann, Lars Jurzik, Ibrahim Ahmed Hamza, Johannes Pinnekamp
ABSTRACT
Fecal contamination of water resources is a major public health concern in densely populated areas since these water bodies are used for drinking water production or recreational purposes. A main source of this contamination originates from combined sewer overflows (CSOs) in regions with combined sewer systems. Thus, the treatment of CSO discharges is urgent. In this study, we explored whether ozonation or UV irradiation can efficiently reduce pathogenic bacteria, viruses, and protozoan parasites in CSOs. Experiments were carried out in parallel settings at the outflow of a stormwater settling tank in the Ruhr area, Germany. The results showed that both techniques reduce most hygienically relevant bacteria, parasites and viruses. Under the conditions tested, ozonation yielded lower outflow values for the majority of the tested parameters.
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
Indole, ≥99%
Sigma-Aldrich
Indole, ≥99%, FG
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%