Merck
  • Evidence That Chlorinated Auxin Is Restricted to the Fabaceae But Not to the Fabeae.
Evidence That Chlorinated Auxin Is Restricted to the Fabaceae But Not to the Fabeae.
Plant physiology (2015-05-15)
Hong Kiat Lam, Scott A M McAdam, Erin L McAdam, John J Ross
ABSTRACT
Auxin is a pivotal plant hormone, usually occurring in the form of indole-3-acetic acid (IAA). However, in maturing pea (Pisum sativum) seeds, the level of the chlorinated auxin, 4-chloroindole-3-acetic acid (4-Cl-IAA), greatly exceeds that of IAA. A key issue is how plants produce halogenated compounds such as 4-Cl-IAA. To better understand this topic, we investigated the distribution of the chlorinated auxin. We show for the first time, to our knowledge, that 4-Cl-IAA is found in the seeds of Medicago truncatula, Melilotus indicus, and three species of Trifolium. Furthermore, we found no evidence that Pinus spp. synthesize 4-Cl-IAA in seeds, contrary to a previous report. The evidence indicates a single evolutionary origin of 4-Cl-IAA synthesis in the Fabaceae, which may provide an ideal model system to further investigate the action and activity of halogenating enzymes in plants.
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
SAFC
Glycine
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Gibberellin, 80% gibberellin A3 basis (TLC)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Gibberellic acid, BioReagent, suitable for plant cell culture, ≥90% gibberellin A3 basis (of total gibberellins.)
Sigma-Aldrich
Gibberellic acid, 90% gibberellin A3 basis (HPLC)