Merck
  • Full-Thickness Astigmatic Keratotomy Combined With Small-Incision Lenticule Extraction to Treat High-Level and Mixed Astigmatism.
Full-Thickness Astigmatic Keratotomy Combined With Small-Incision Lenticule Extraction to Treat High-Level and Mixed Astigmatism.
Cornea (2015-09-30)
Bu Ki Kim, Su Joung Mun, Dae Gyu Lee, Jae Ryun Kim, Hyun Seung Kim, Young Taek Chung
ABSTRACT
To explore the clinical effects of combined full-thickness astigmatic keratotomy and small-incision lenticule extraction (SMILE) in patients who are inoperable using SMILE alone. We included 13 eyes of 9 patients with high-level or mixed astigmatism who underwent full-thickness astigmatic keratotomy followed by SMILE (secondarily) to correct the residual refractive error. Six months after SMILE, the spherical equivalent was reduced from -4.83 ± 3.26 D to -0.17 ± 0.38 D (P < 0.001), and the astigmatism was reduced from 5.12 ± 0.96 D to 0.21 ± 0.22 D (P < 0.001). The uncorrected and corrected (CDVA) distance visual acuities improved from 1.07 ± 0.62 to 0.02 ± 0.13 (P < 0.001) and from 0.08 ± 0.14 to -0.01 ± 0.14 (P = 0.002), respectively. The CDVA improved by 1 or 2 Snellen lines in 8 cases (61.5%), and there was no loss in CDVA. All procedures were completed without intraoperative or postoperative complications. This combined procedure was effective and safe for the treatment of high-level or mixed astigmatism.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Fluorometholone, ≥98%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid