Merck
  • Moderate-intensity exercise training elevates serum and pancreatic zinc levels and pancreatic ZnT8 expression in streptozotocin-induced diabetic rats.
Moderate-intensity exercise training elevates serum and pancreatic zinc levels and pancreatic ZnT8 expression in streptozotocin-induced diabetic rats.
Life sciences (2015-08-19)
Juraiporn Somboonwong, Supitchaya Traisaeng, Sompol Saguanrungsirikul
ABSTRACT
Diabetes mellitus is associated with disturbed zinc homeostasis and down-regulation of zinc transporter 8 (ZnT8); these changes contribute to the defective biosynthesis, storage, and secretion of insulin. Previous studies have reported an improvement in diabetic status and insulin levels in diabetic rats that underwent moderate exercise training, but the mechanisms underlying this effect remain unclear. Evidence shows that exercise training increases the zinc content in the muscle, liver, and kidney of diabetic rats and increases the expression of several types of zinc transporters in the rat hippocampus. We hypothesised that moderate exercise training may increase serum and pancreatic zinc levels, as well as pancreatic ZnT8 expression, in diabetic rats. Wistar rats were divided into 3 equally sized groups: sedentary normal control, sedentary diabetic, and exercise-trained diabetic groups. Diabetes was induced by an intraperitoneal injection of streptozotocin. The 6-week exercise training intervention involved 30 min of moderate-intensity running on a treadmill once daily (5 days/week). At the end of the study, the concentrations of serum and pancreatic zinc were determined using atomic absorptive spectrophotometry. Pancreatic ZnT8 expression was analysed by quantitative real-time RT-PCR. Diabetes caused reductions in the serum and pancreatic zinc levels and pancreatic ZnT8 expression. Following moderate exercise training, there was a significant increase in all of these parameters. The ability of moderate exercise training to ameliorate the reductions in serum and pancreatic zinc levels and pancreatic ZnT8 expression can partly explain the beneficial effects of exercise training in diabetes.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Citric acid trisodium salt, ≥98% (GC), anhydrous
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
3-Methyl-1-butanol, reagent grade, 98%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%