Merck
  • Cuprizone Model as a Tool for Preclinical Studies of the Efficacy of Multiple Sclerosis Diagnosis and Therapy.
Cuprizone Model as a Tool for Preclinical Studies of the Efficacy of Multiple Sclerosis Diagnosis and Therapy.
Bulletin of experimental biology and medicine (2015-06-02)
T O Abakumova, A A Kuz'kina, M E Zharova, M V Zharova, D A Pozdeeva, I L Gubskii, I I Shepeleva, O M Antonova, N V Nukolova, Z I Kekelidze, V P Chekhonin
ABSTRACT
To study demyelination and remyelination processes and their response to different drugs, a protocol for modeling multiple sclerosis using the copper chelator cuprizone was developed. Magnetic resonance imaging confirmed the presence of demyelination lesions on week 4 of 0.6% cuprizone-containing diet. Immunohistochemical staining with polyclonal antibodies to glial fibrillary acidic protein (pAb GFAP) confirmed the increase in the number of reactive astrocytes on week 4 of diet and during remyelination (week 2 after diet). Analysis of neurophysiological functions in mice with cuprizone-induced demyelination revealed motor and behavioral deficits. This model can be used as a tool for preclinical studies of the efficiency of multiple sclerosis diagnostic and therapy.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%