Merck
  • Helix pomatia hemocyanin - a novel bio-adjuvant for viral and bacterial antigens.
Helix pomatia hemocyanin - a novel bio-adjuvant for viral and bacterial antigens.
International immunopharmacology (2015-03-25)
Vera Gesheva, Stela Chausheva, Nadia Stefanova, Nikolina Mihaylova, Lyuba Doumanova, Krassimira Idakieva, Andrey Tchorbanov
ABSTRACT
New generated subunit vaccines are characterized by increased safety and lack of side effects, however they suffer from weak immunogenicity. The adjuvants are substances that have the ability to enhance the magnitude and duration of the immune response and to increase vaccine efficacy, but the different vaccines may require diverse adjuvants. The urgent need of novel adjuvant formulations occurs, thus ensuring protective cellular and humoral responses against infectious pathogens. The hemocyanins, oxygen binding copper proteins in the hemolymph of molluscs and arthropods, are widely used as peptide carriers and vaccine adjuvants. In the present study we promote the hemocyanin isolated from the terrestrial gastropod Helix pomatia (HPH) as bio-adjuvant, combined with standard antigens. The purified HPH combined with influenza virus hemagglutinin intersubunit peptide (IP) or with tetanus toxoid (TT) were used for immunization. Administration of tetanus toxoid combined with HPH in mice resulted in an increased number of anti-TT IgG producing plasmocytes and induced a significant increase of B and T cell proliferation. The level of the anti-TT IgG antibodies in mice sera was comparable to the group administered with TT+Al(OH)3. An immunization of experimental animals with IP combined with H. pomatia hemocyanin led to generation of strong anti-influenza cytotoxic response. The vaccination of mice demonstrates that the HPH is acceptable as a potential bio-adjuvant for subunit vaccines and it could be used as a natural adjuvant or protein carrier.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O