Merck
  • Concentrations of Contaminants with Regulatory Limits in Samples of Clam (Chamelea gallina) Collected along the Abruzzi Region Coast in Central Italy.
Concentrations of Contaminants with Regulatory Limits in Samples of Clam (Chamelea gallina) Collected along the Abruzzi Region Coast in Central Italy.
Journal of food protection (2015-09-01)
Pierina Visciano, Giampiero Scortichini, Giovanna Suzzi, Gianfranco Diletti, Maria Schirone, Giuseppe Martino
ABSTRACT
Concentrations of pollutants with regulatory limits were determined in specimens of Chamelea gallina, a species of clam collected along the Abruzzi coastal region of the central Adriatic Sea. Nine sampling sites were selected to evaluate the distribution of contaminants in the environment and the health risk for consumers. The concentrations of all the examined compounds were lower than the maximums set by European legislation. Polycyclic aromatic hydrocarbons and total mercury were below the detection limit (0.18 μg/kg for benzo[a]anthracene, 0.30 μg/kg for chrysene, 0.12 μg/kg for benzo[b]fluoranthene, 0.08 μg/kg for benzo[a]pyrene, and 0.0050 mg/kg for total mercury) in all the analyzed samples. Mean concentrations of lead and cadmium were 0.104 and 0.110 mg/kg, respectively. Of the non-dioxin-like polychlorinated biphenyls, PCB-153, PCB-180, and PCB-138 were the most abundant at all sampling sites (1a to 9a) at 0.25 mi (ca. 0.4 km) and at some sampling sites (1b, 2b, 3b, 5b and 7b) at 0.35 mi (ca. 0.56 km). Principal component analysis revealed that the concentrations of dioxin-like polychlorinated biphenyls were similar at the majority of sampling sites, and O8CDD and 2,3,7,8-T4CDF were the predominant dioxin congeners.
Sigma-Aldrich
2,2,4-Trimethylpentane, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Rhenium, powder, −100 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Rhenium, powder, 99.995% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%