Merck
  • Cetylpyridinium chloride and miramistin as antiseptic substances in chronic wound management - prospects and limitations.
Cetylpyridinium chloride and miramistin as antiseptic substances in chronic wound management - prospects and limitations.
Journal of medical microbiology (2015-02-15)
Carolin Fromm-Dornieden, Julian-Dario Rembe, Nadine Schäfer, Julia Böhm, Ewa K Stuermer
ABSTRACT
The antimicrobial activity of cetylpyridinium chloride (CPC) and miramistin (MST) solutions at different concentrations (5×10(-5) to 0.4%) and a dressing, containing 0.15% CPC, were tested against Staphylococcus aureus, Pseudomonas aeruginosa and Escherichia coli after 30 (solutions) and 60 min (fleece) incubation, respectively. Furthermore, the cytotoxic effects of CPC and MST were examined in human keratinocyte (HaCaT) and murine fibroblast (L929) cell lines. A dose of 3×10(-3)% CPC or MST was sufficient to entirely eradicate S. aureus after 30 min incubation. To achieve the same effect, higher concentrations were required against E. coli (0.025% CPC; 0.0125% MST) and P. aeruginosa (0.5% CPC; 0.05% MST). The CPC-fleece showed a high antiseptic effect against all three bacterial strains, although it did not completely eliminate P. aeruginosa. Both substances showed a high cytotoxic impact at higher tested concentrations (CPC >3×10(-3)%; MST >8×10(-4)%). CPC showed high antimicrobial potency at low concentrations against S. aureus, accompanied by low cytotoxic (side) effects at these concentrations, whilst the required minimal concentration to eradicate E. coli and P. aeruginosa was shown to be cytotoxic for keratinocytes and fibroblasts. The necessary antibacterial amounts of MST were lower, but also cytotoxic in direct contact with typical human wound cells. With regard to demographic changes and increasing bacterial resistance, new effective antiseptics, such as CPC and MST, incorporated in wound dressings without releasing an active substance could help to improve the treatment and healing rates of chronic wounds.
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O