Merck
  • Idiotypic DNA vaccination for the treatment of multiple myeloma: safety and immunogenicity in a phase I clinical study.
Idiotypic DNA vaccination for the treatment of multiple myeloma: safety and immunogenicity in a phase I clinical study.
Cancer immunology, immunotherapy : CII (2015-05-20)
Katy J McCann, Rosemary Godeseth, Lindsey Chudley, Ann Mander, Gianfranco Di Genova, Paul Lloyd-Evans, Jonathan P Kerr, Vladimir B Malykh, Matthew W Jenner, Kim H Orchard, Freda K Stevenson, Christian H Ottensmeier
ABSTRACT
We report on the safety and immunogenicity of idiotypic DNA vaccination in a phase I, non-randomised, open-label study in patients with multiple myeloma. The study used DNA fusion gene vaccines encoding patient-specific single chain variable fragment, or idiotype (Id), linked to fragment C (FrC) of tetanus toxin. Patients in complete or partial response following high-dose chemotherapy and autologous stem cell transplant were vaccinated intramuscularly with 1 mg DNA on six occasions, beginning at least 6 months post-transplant; follow-up was to week 52. Fourteen patients were enrolled on study and completed vaccinations. Idiotypic DNA vaccines were well tolerated with vaccine-related adverse events limited to low-grade constitutional symptoms. FrC- and Id-specific T-cell responses were detected by ex vivo ELISPOT in 9/14 and 3/14 patients, respectively. A boost of pre-existing anti-FrC antibody (Ab) was detected by ELISA in 8/14 patients, whilst anti-Id Ab was generated in 1/13 patients. Overall, four patients (29 %) made an immune response to FrC and Id, with six patients (43 %) responding to FrC alone. Over the 52-week study period, serum paraprotein was undetectable, decreased or remained stable for ten patients (71 %), whilst ongoing CR/PR was maintained for 11 patients (79 %). The median time to progression was 38.0 months for 13/14 patients. Overall survival was 64 % after a median follow-up of 85.6 months.
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Vinyl laurate, Wacker Chemie AG, ≥98% (GC)
Sigma-Aldrich
Vinyl laurate, ≥99.0% (GC)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Supelco
N,N′-Bis(acryloyl)cystamine, BioReagent, suitable for electrophoresis
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
SAFC
BIS-TRIS
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sigma-Aldrich
Ammonium hydrogen difluoride, 99.999% trace metals basis