Merck
  • A Standardized Extract of Rhus verniciflua Stokes Protects Wistar Rats Against Lipopolysaccharide-Induced Acute Inflammation.
A Standardized Extract of Rhus verniciflua Stokes Protects Wistar Rats Against Lipopolysaccharide-Induced Acute Inflammation.
Journal of medicinal food (2015-10-27)
Ji Eun Moon, Jae-Ho Shin, Oran Kwon, Ji Yeon Kim
ABSTRACT
Rhus verniciflua stokes (RVS) (Anacardiaceae) has been traditionally used as a folk remedy for gastritis, several cancers, and various metabolic diseases. The present study evaluated the anti-inflammatory effect of RVS extract standardized to fustin content using lipopolysaccharide (LPS)-stimulated rats. The rats were randomly divided into six groups and intragastrically administered 0, 100, 250, or 500 mg/kg body weight (bw) of RVS or 15 mg/kg bw of fustin for 14 days. LPS was intraperitoneally injected 18 h before sacrifice. The nitric oxide levels of RVS extract in either the serum or liver were significantly decreased compared to the LPS-treated rats (P<.05). The treatment with the RVS extract also blunted the rise of malondialdehyde levels in the liver (P<.05). The administration of RVS extract and fustin significantly prevented the elevation of interleukin 6 cytokine, iNOS, and COX-2 mRNA expression in the liver. Inflammatory cell infiltration was also significantly attenuated by the RVS extract or fustin supplementation. These results suggest that our standardized RVS extract has preventive effects on inflammatory reactions.
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS reagent, >98%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
N-(1-Naphthyl)ethylenediamine dihydrochloride, ≥98%