Merck
  • Predictors of heroin relapse: Personality traits, impulsivity, COMT gene Val158met polymorphism in a 5-year prospective study in Shanghai, China.
Predictors of heroin relapse: Personality traits, impulsivity, COMT gene Val158met polymorphism in a 5-year prospective study in Shanghai, China.
American journal of medical genetics. Part B, Neuropsychiatric genetics : the official publication of the International Society of Psychiatric Genetics (2015-09-09)
Hang Su, Zhibin Li, Jiang Du, Haifeng Jiang, Zhikang Chen, Haiming Sun, Min Zhao
ABSTRACT
Relapse is a typical feature of heroin addiction and rooted in genetic and psychological determinants. The aim of this study was to evaluate the effect of personality traits, impulsivity, and COMT gene polymorphism (rs4680) on relapse to heroin use during 5-year follow up. 564 heroin dependent patients were enrolled in compulsory drug rehabilitation center. 12 months prior to their release, personality traits were measured by BIS-11 (Barratt Impulsiveness Scale-11) and Temperament and Character Inventory (TCI). The COMT gene rs4680 polymorphism was genotyped using a DNA sequence detection system. The heroin use status was evaluated for 5 years after discharged. Among the 564 heroin-dependent patients, 500 were followed for 5 years after discharge and 53.0% (n = 265) were considered as relapsed to heroin use according to a strict monitor system. Univariate analysis showed that age, having ever been in methadone maintenance treatment (MMT), the total scores and non-planning scores of BIS-11, and the COMT rs4680 gene variants were different between relapse and abstinent groups. Logistic regression analysis showed higher BIS total score, having ever been in MMT and younger first heroin use age are the predictors of relapse to heroin use during 5 years follow-up, and the COMT rs4680 gene had an interaction with BIS scores. Our findings indicated that the impulsive personality traits, methadone use history, and onset age could predict relapse in heroin-dependent patients during 5 year's follow up. The COMT gene showed a moderational effect in part the relationship of impulsivity with heroin relapse.
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O