Merck
  • Detection of ethanolamine altering in fetuses of pregnancy-associated hypertensive mice treated with vasodepressors by using UPLC and MALDI-TOF/MS.
Detection of ethanolamine altering in fetuses of pregnancy-associated hypertensive mice treated with vasodepressors by using UPLC and MALDI-TOF/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-11-04)
Koichiro Kako, Ayumi Nakamura, Yusuke Nagashima, Junji Ishida, Akiyoshi Fukamizu
ABSTRACT
Since serotonin, homocysteine and oxytocin are known to fluctuate during mammalian gestation, we screened amines altered in pregnant-associated hypertensive (PAH) mice by tagging their amino groups with 6-aminoquinoline carbamoyl (AQC) group in concert with ultra high-performance liquid chromatography (UPLC). Interestingly, a candidate amine significantly increased in PAH mice was recovered to the basal level, when treated with antihypertensive drugs. Mass spectrometric analyses indicated that the molecular mass of this amine was 61.2, which was identified as ethanolamine.
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
L-Norvaline, arginase inhibitor
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%