Merck
  • Periconceptional undernutrition modifies endocrine profiles and hepatic gene expression in sheep.
Periconceptional undernutrition modifies endocrine profiles and hepatic gene expression in sheep.
Journal of animal physiology and animal nutrition (2014-10-17)
V de Brun, A Meikle, A Casal, M Sequeira, I Contreras-Solís, M Carriquiry, F Forcada, C Sosa, J A Abecia
ABSTRACT
This study investigated whether a 22-day period of undernutrition (half maintenance) could affect maternal endocrine responses and liver gene expression during early pregnancy (day 7). Thirty-five ewes were fed 1.5 (n = 15) or 0.5 (n = 20) their maintenance requirements and slaughtered on day 7 of the oestrus cycle or pregnancy (oestrus = day 0). Insulin, IGF, leptin and non-esterified fatty acids (NEFA) were determined on days -14, 0 and 7. Transcripts of the IGF family and adipokines receptors were determined in the liver by real-time RT-PCR. Underfed animals presented lower body weight and body condition, greater plasma concentration of NEFA, and lower plasma concentrations of leptin, insulin and IGF1 compared to adequately fed animals. Underfed ewes presented greater hepatic expression of IGFBP2 than well-fed ewes, but tended to have lesser expression of IGFBP5. While no effect of undernutrition on IGFBP4 and ADIPOR2 mRNA expressions was observed, they were increased by pregnancy in underfed animals. This study shows that undernutrition modifies endocrine profiles and hepatic gene expression of IGFBP2 and 5. The pregnancy status increased hepatic gene expression of IGFBP4 and ADIPOR2 mRNA in undernourished ewes.
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)