Merck
  • Haplotype distribution in the class I sirtuin genes and their associations with ultrasound carcass traits in Qinchuan cattle (Bos taurus).
Haplotype distribution in the class I sirtuin genes and their associations with ultrasound carcass traits in Qinchuan cattle (Bos taurus).
Molecular and cellular probes (2015-04-04)
Linsheng Gui, Ruijie Hao, Yaran Zhang, Xianlin Zhao, Linsen Zan
ABSTRACT
Class I sirtuin genes including SIRT1, SIRT2 and SIRT3, are members of the nicotinamide adenine dinucleotide (NAD)-dependent family of histone deacetylases, and play essential roles in senescence, metabolism, and apoptosis. This study was conducted to detect potential polymorphisms of the bovine class I sirtuin genes and explore their relationships with ultrasound carcass traits in Qinchuan cattle. Four non-coding mutations in the 3'UTR (SIRT1: g.25751A > C, SIRT1: g.25846A > G, SIRT2: g.19676G > A and SIRT3: g. 25702C > T) and three mutations in exons (SIRT2: g.4062C > T; SIRT2: g.4406C > T and SIRT3: g.25557A > G) were identified in 468 individuals of Qinchuan cattle. Chi-square tests showed that g.25751A > C, g.19676G > A, and g.25702C > T were in Hardy-Weinberg disequilibrium (χ(2) < χ0.05(2)). The statistical analyses indicated that six SNPs were significantly associated with the ultrasound carcass traits (P < 0.05) except g.4062C > T (SIRT2) (P > 0.05). These results indicate that the variations in the class I sirtuin genes and their corresponding genotypes may be considered as molecular markers for economic traits in cattle breeding.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O