Merck
  • Quinopeptide formation associated with the disruptive effect of epigallocatechin-gallate on lysozyme fibrils.
Quinopeptide formation associated with the disruptive effect of epigallocatechin-gallate on lysozyme fibrils.
International journal of biological macromolecules (2015-05-02)
Na Cao, Yu-Jie Zhang, Shuang Feng, Cheng-Ming Zeng
ABSTRACT
Numerous studies demonstrate that natural polyphenols can inhibit amyloid formation and disrupt preformed amyloid fibrils. In the present study, the fibril-disruptive effects of epigallocatechin-3-gallate (EGCG) were examined using lysozyme as a model protein. The results indicated that EGCG dose dependently inhibited lysozyme fibrillation and modified the peptide chains with quinonoid moieties under acidic conditions, as measured by ThT fluorescence, transmission electron microscopy, and an NBT-staining assay. Moreover, EGCG transformed the preformed lysozyme fibrils to amorphous aggregates through quinopeptide formation. The thiol blocker, N-ethylmaleimide, inhibited the disruptive effect of EGCG on preformed fibrils, suggesting that thiol groups are the binding sites for EGCG. We propose that the formation of quinone intermediates via oxidation and subsequent binding to lysozyme chains are the main processes driving the inhibition of amyloid formation and disruption of preformed fibrils by EGCG. The information presented in this study may provide fresh insight into the link between the antioxidant capacity and anti-amyloid activity of polyphenols.
Sigma-Aldrich
N-Ethylmaleimide, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade