Merck
  • Estimation of human leptin concentration in the subcutaneous adipose and skeletal muscle tissues.
Estimation of human leptin concentration in the subcutaneous adipose and skeletal muscle tissues.
European journal of clinical investigation (2015-02-25)
Gerald Sendlhofer, Gernot Brunner, Lukas Schaupp, Andrea Wutte, Martin Ellmerer, Thomas R Pieber
ABSTRACT
Interstitial leptin concentrations in subcutaneous adipose and skeletal muscle tissues were determined by open-flow microperfusion. In 12 lean male subjects (age: 25.6 ± 1.1 years), a zero flow rate experiment using different flow rates was applied. Recovery was determined by urea as an internal reference. In the no-net-flux experiments, catheters were perfused with five solutions containing different concentrations of leptin. Concentrations of interstitial leptin were calculated by applying linear regression analysis to perfusate as opposed to sampled leptin concentrations. The zero flow rate protocol showed significantly higher concentrations of leptin in the interstitial fluid of subcutaneous adipose compared to skeletal muscle tissue [36.8 ± 10.32 vs. 7.1 ± 2.5% of the corresponding plasma level (P = 0.018)]. The recovery of urea in the samples was comparable for all catheters [79.4 ± 6.8 vs. 83.0 ± 5.8 of the corresponding plasma level, flow rate of 0.3 μL/min; (P = ns)] and was higher when compared to leptin. In the no-net-flux protocol, the concentration of leptin in subcutaneous adipose tissue was almost identical to plasma [90. 5 ± 7.0%] and the skeletal muscle tissue concentration of leptin was 23.7 ± 2.5% of the corresponding plasma level. Open-flow microperfusion enables the estimation of leptin concentrations in subcutaneous adipose and skeletal muscle tissues in humans in vivo. This is the first documentation on the use of open-flow microperfusion to demonstrate that relevant amounts of leptin are also found in skeletal muscle tissue.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)