Merck
  • Preparation of an engineered safer immunotoxin against colon carcinoma based on the ribotoxin hirsutellin A.
Preparation of an engineered safer immunotoxin against colon carcinoma based on the ribotoxin hirsutellin A.
The FEBS journal (2015-03-11)
Jaime Tomé-Amat, Elías Herrero-Galán, Mercedes Oñaderra, Álvaro Martínez-Del-Pozo, José G Gavilanes, Javier Lacadena
ABSTRACT
Immunotoxins are chimeric proteins composed of an antibody domain that specifically directs the action of the toxic domain, resulting in the death of the targeted cells. Over recent years, immunotoxins have been widely studied and the number of different constructions has increased exponentially. Protein engineering has allowed the design of optimized versions of immunotoxins with an improved tumor binding affinity, stability or cytotoxic efficacy, although sometimes this has compromised the safety of the patient in terms of undesirable adverse secondary reactions. A triple mutant at three Trp residues (HtA3ΔW) of the ribotoxin hirsutellin A retains its specific ribonucleolytic activity, although cell internalization capacity is lacking. This toxin variant has been fused to the single chain variable fragment A33 (scFvA33). This immunoconjugate (IMTXA33HtA3ΔW) was produced in the methylotrophic yeast Pichia pastoris and purified using nickel-nitrilotriacetic acid affinity chromatography. Both target and toxic domains were characterized. The immunotoxin showed an exquisite specific binding against GPA33-positive culture cells, which results in the death of the targeted cells because of specific ribonucleolytic activity against ribosomes of the engineered hirsutellin A variant. IMTXA33HtA3ΔW represents a promising structure in the search for an improved immunotoxin without compromising the safety of patients.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Nitrilotriacetic acid, ACS reagent, ≥99.0%
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Nitrilotriacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)