Merck
  • Crescendo: A Protein Sequence Database Search Engine for Tandem Mass Spectra.
Crescendo: A Protein Sequence Database Search Engine for Tandem Mass Spectra.
Journal of the American Society for Mass Spectrometry (2015-04-22)
Jianqi Wang, Yajie Zhang, Yonghao Yu
ABSTRACT
A search engine that discovers more peptides reliably is essential to the progress of the computational proteomics. We propose two new scoring functions (L- and P-scores), which aim to capture similar characteristics of a peptide-spectrum match (PSM) as Sequest and Comet do. Crescendo, introduced here, is a software program that implements these two scores for peptide identification. We applied Crescendo to test datasets and compared its performance with widely used search engines, including Mascot, Sequest, and Comet. The results indicate that Crescendo identifies a similar or larger number of peptides at various predefined false discovery rates (FDR). Importantly, it also provides a better separation between the true and decoy PSMs, warranting the future development of a companion post-processing filtering algorithm.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Universal Proteomics Standard Set, Protein Mass Spectrometry Calibration Standard
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
SAFC
Iodoacetamide