Merck
  • Phenolic compounds in grains, sprouts and wheatgrass of hulled and non-hulled wheat species.
Phenolic compounds in grains, sprouts and wheatgrass of hulled and non-hulled wheat species.
Journal of the science of food and agriculture (2014-08-19)
Paolo Benincasa, Angelica Galieni, Anna Chiara Manetta, Roberta Pace, Marcello Guiducci, Michele Pisante, Fabio Stagnari
ABSTRACT
The use of sprouts and young plantlets in human nutrition is increasing because they often contain phytochemicals and other high value nutrients. This is also the case for wheat, although there is no literature for hulled wheat species. Thus we determined total polyphenols, phenolic acids (PAs), fibre and minerals in grains, 5-day-old sprouts and 12-day-old wheatgrass of einkorn (cv. Monlis), emmer (cvs Augeo, Rosso Rubino, Zefiro), spelt (cvs Pietro, Giuseppe), durum wheat (cv. Creso) and soft wheat (cv. Orso). Grains of einkorn and emmer contained twice bound PAs as compared to soft and durum wheat and spelt, with p-coumaric acid accounting for about 50% of total bound PAs. In wheatgrass, differences between species for bound PAs decreased due to a decrease in einkorn and emmer and an increase in soft and durum wheat. In all species, total phenols and free PAs increased passing from grains to sprouts and wheatgrass. Neutral and acid detergent fibre content increased with sprouting only in einkorn and emmer. Our evidence suggests that the grains of einkorn and emmer and the sprouts and wheatgrass of all Triticum species might potentially be valuable for the development of functional foods.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Potassium hydride, in paraffin