Merck
  • VEGF Enhances the Migration of MSCs in Neural Differentiation by Regulating Focal Adhesion Turnover.
VEGF Enhances the Migration of MSCs in Neural Differentiation by Regulating Focal Adhesion Turnover.
Journal of cellular physiology (2015-03-31)
Huihui Wang, Xingkai Wang, Jing Qu, Qing Yue, Ya'nan Hu, Huanxiang Zhang
ABSTRACT
Mesenchymal stem cells (MSCs) hold great promise in neural regeneration, due to their intrinsic neuronal potential and migratory tropism to damaged nervous tissues. However, the chemotactic signals mediating the migration of MSCs remain poorly understood. Here, we investigated the regulatory roles for focal adhesion kinase (FAK) and Rac1 in vascular endothelial growth factor (VEGF)-stimulated migration of MSCs in neural differentiation. We found that MSCs in various differentiation states show significant different chemotactic responses to VEGF and cells in 24-h preinduction state possess the highest migration speed and efficiency. FAK, as the downstream signaling molecule, is involved in the VEGF-induced migration by regulating the assembly and distribution of focal adhesions (FAs) and reorganization of F-actin. The features of FAs and cytoskeletons and the ability of lamellipodia formation are closely related to the neural differentiation states of MSCs. VEGF promotes FA formation with an asymmetric distribution of FAs and induces the activation of Y397-FAK and Y31/118-paxillin of undifferentiated and 24-h preinduced MSCs in a time-dependent manner. Inhibition of FAK by PF-228 or expressing FAK-Y397F mutant impairs the dynamics of FAs in MSCs during VEGF-induced migration. Furthermore, Rac1 regulates FA formation in a FAK-dependent manner. Overexpression of constitutive activated mutants of Rac1 increases the number of FAs in undifferentiated and 24-h preinduced MSCs, while VEGF-induced increase of FA formation is decreased by inhibiting FAK by PF-228. Collectively, these results demonstrate that FAK and Rac1 signalings coordinately regulate the dynamics of FAs during VEGF-induced migration of MSCs in varying neural differentiation states.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis