Merck
  • Thr175-phosphorylated tau induces pathologic fibril formation via GSK3β-mediated phosphorylation of Thr231 in vitro.
Thr175-phosphorylated tau induces pathologic fibril formation via GSK3β-mediated phosphorylation of Thr231 in vitro.
Neurobiology of aging (2015-01-13)
Alexander J Moszczynski, May Gohar, Kathryn Volkening, Cheryl Leystra-Lantz, Wendy Strong, Michael J Strong
ABSTRACT
We have previously shown that amyotrophic lateral sclerosis with cognitive impairment can be characterized by pathologic inclusions of microtubule-associated protein tau (tau) phosphorylated at Thr(175) (pThr(175)) in association with GSK3β activation. We have now examined whether pThr(175) induces GSK3β activation and whether this leads to pathologic fibril formation through Thr(231) phosphorylation. Seventy-two hours after transfection of Neuro2A cells with pseudophosphorylated green fluorescent protein-tagged 2N4R tau (Thr(175)Asp), phosphorylated kinase glycogen synthase kinase 3 beta (active GSK3β) levels were significantly increased as was pathologic fibril formation and cell death. Treatment with each of 4 GSK3β inhibitors or small hairpin RNA knockdown of GSK3β abolished fibril formation and prevented cell death. Inhibition of Thr(231) phosphorylation (Thr(231)Ala) prevented pathologic tau fibril formation, regardless of Thr(175) state, whereas Thr(231)Asp (pseudophosphorylated at Thr(231)) developed pathologic tau fibrils. Ser(235) mutations did not affect fibril formation, indicating an unprimed mechanism of Thr(231) phosphorylation. These findings suggest a mechanism of tau pathology by which pThr(175) induces GSK3β phosphorylation of Thr(231) leading to fibril formation, indicating a potential therapeutic avenue for amyotrophic lateral sclerosis with cognitive impairment.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
AR-A014418, ≥98% (HPLC), solid
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%