Merck
  • An amplified electrochemical strategy using DNA-QDs dendrimer superstructure for the detection of thymine DNA glycosylase activity.
An amplified electrochemical strategy using DNA-QDs dendrimer superstructure for the detection of thymine DNA glycosylase activity.
Biosensors & bioelectronics (2015-04-29)
Hongying Liu, Youbing Lou, Fei Zhou, Hao Zhu, E S Abdel-Halim, Jun-Jie Zhu
ABSTRACT
A triple-signal amplification strategy was proposed for highly sensitive and selective detection of thymine DNA glycosylase (TDG) by coupling a dendrimer-like DNA label with the electrochemical method and quantum dots (QDs) tagging. The DNA-QDs dendrimer-like superstructure was designed by DNA hybridization and covalent assembling. Benefiting from outstanding performance of the amplification strategy, this assay showed high sensitivity, extraordinary stability, and easy operation. The limit of detection could reach 0.00003 U µL(-1) with a splendid specificity. The TDG content in different concentration of HeLa cell was also determined. This assay opens a new horizon for both qualitative and quantitative detection of TDG, holding great promise for potential application in cancer cell research and clinical diagnostics.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
1-Hexanethiol, 99% (GC)
Sigma-Aldrich
1-Hexanethiol, 95%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
6-Mercapto-1-hexanol, 97%
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Thymine, ≥99%
Sigma-Aldrich
Thymine, BioReagent, suitable for cell culture
Sigma-Aldrich
6-Mercapto-1-hexanol, 99%
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99%
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99.0% (HPLC)
Sigma-Aldrich
Trizma® base, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Phosphorus, red, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%