Merck
  • Metabolomic fingerprinting of saffron by LC/MS: novel authenticity markers.
Metabolomic fingerprinting of saffron by LC/MS: novel authenticity markers.
Analytical and bioanalytical chemistry (2015-07-23)
Miguel Guijarro-Díez, Leonor Nozal, María Luisa Marina, Antonio Luis Crego
ABSTRACT
An untargeted metabolomic approach using liquid chromatography coupled to electrospray ionization time-of-flight mass spectrometry was developed in this work to identify novel markers for saffron authenticity which is an important matter related to consumer protection, quality assurance, active properties, and also economical impact (saffron is the most expensive spice). Metabolic fingerprinting of authentic and suspicious saffron samples from different geographical origin was obtained and analyzed. Different extracting protocols and chromatographic methodologies were evaluated to obtain the most adequate extracting and separation conditions. Using an ethanol/water mixture at pH 9.0 and an elution gradient with a fused core C18 column enabled obtaining the highest number of significant components between authentic and adulterated saffron. By using multivariate statistical analysis, predictive classification models for authenticity and geographical origin were obtained. Moreover, 84 and 29 significant metabolites were detected as candidates for markers of authenticity and geographical origin, respectively, from which only 34 metabolites were tentatively identified as authenticity markers of saffron, but none related to its geographical origin. Six characteristic compounds of saffron (kaempferol 3-O-glucoside, kaempferol 3-O-sophoroside, kaempferol 3,7-O-diglucoside, kaempferol 3,7,4'-O-triglucoside, kaempferol 3-O-sophoroside-7-O-glucoside, and geranyl-O-glucoside) were confirmed by comparing experimental MS/MS fragmentation patterns with those provided in scientific literature being proposed as novel markers of authenticity. Graphical Abstract Metabolomic fingerprinting of saffron.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Geranic acid, 85%, stabilised
Sigma-Aldrich
Geranic acid, technical grade, 85%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Baicalein, 98%
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Terfenadine
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Niflumic acid
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis