Merck
  • Determination of four paraben-type preservatives and three benzophenone-type ultraviolet light filters in seafoods by LC-QqLIT-MS/MS.
Determination of four paraben-type preservatives and three benzophenone-type ultraviolet light filters in seafoods by LC-QqLIT-MS/MS.
Food chemistry (2015-10-17)
Chao Han, Biqi Xia, Xiangzhun Chen, Jincan Shen, Qian Miao, Yan Shen
ABSTRACT
For the first time, an efficient and sensitive analytical method based on liquid chromatography-quadrupole linear ion trap-tandem mass spectrometry (LC-QqLIT-MS/MS) was developed for the simultaneous determination of four paraben-type preservatives and three benzophenone-type ultraviolet light filters in both plant (Sargassum fusiforme, porphyra, kelp) and animal (hairtail, yellow croaker, shrimp) seafood. The samples were extracted in methanol by pressurized liquid extraction (PLE), and the extracts were then cleaned up by mixed-mode cationic exchange (MCX) solid-phase extraction cartridges. Both isotope-labeled internal standards and matrix-matched calibration standards were used to alleviate and correct for the matrix effects, and the limits of quantification (LOQs) were 10.0μg kg(-1) for all target compounds. The average recoveries were in the range of 80.6-107.8% at three spiked concentration levels (10, 50 and 100μgkg(-1)) with relative standard deviations (RSDs) below 8.5%. The results suggest that very limited contamination of these seven emerging contaminants, mainly associated with PCPs, occurred in these common seafoods.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99.0% (GC)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Ethyl 4-hydroxybenzoate, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%