Merck
  • Evaluation of the cytotoxic and antitumour effects of the essential oil from Mentha x villosa and its main compound, rotundifolone.
Evaluation of the cytotoxic and antitumour effects of the essential oil from Mentha x villosa and its main compound, rotundifolone.
The Journal of pharmacy and pharmacology (2015-03-20)
Ricardo G Amaral, Cecília S Fonseca, Tayane Kayne M Silva, Luciana N Andrade, Maria E França, José M Barbosa-Filho, Damião P de Sousa, Manoel O Moraes, Cláudia Ó Pessoa, Adriana A Carvalho, Sara Maria Thomazzi
ABSTRACT
The aim of this study was to investigate the cytotoxic and antitumour effects of the essential oil from the leaves of Mentha x villosa (EOMV) and its main component (rotundifolone). In-vitro cytotoxic activity of the EOMV and rotundifolone was determined on cultured tumour cells. In-vivo antitumour activity of the EOMV was assessed in sarcoma 180-bearing mice. The EOMV displayed cytotoxicity against human tumour cell lines, showing IC50 values in the range of 0.57-1.02 μg/ml in the HCT-116 and SF-295 cell lines, respectively. Rotundifolone showed weak cytotoxicity against HCT-116, SF-295 and OVCAR-8 cell lines (IC50 > 25.00 μg/ml). Tumour growth inhibition rates were 29.4-40.5% and 25.0-45.2% for the EOMV treatment by intraperitoneal (50-100 mg/kg/day) and oral (100-200 mg/kg/day) administration, respectively. The EOMV did not significantly affect body mass and macroscopy of the organs. The EOMV possesses significant antitumour activity with low systemic toxicity, possibly due to the synergistic action of its minor constituents.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®