Merck
  • Mitochondria apoptosis pathway synergistically activated by hierarchical targeted nanoparticles co-delivering siRNA and lonidamine.
Mitochondria apoptosis pathway synergistically activated by hierarchical targeted nanoparticles co-delivering siRNA and lonidamine.
Biomaterials (2015-05-26)
Bing-Feng Zhang, Lei Xing, Peng-Fei Cui, Feng-Zhen Wang, Rong-Lin Xie, Jia-Liang Zhang, Mei Zhang, Yu-Jing He, Jin-Yuan Lyu, Jian-Bin Qiao, Bao-An Chen, Hu-Lin Jiang
ABSTRACT
The mitochondria-mediated apoptosis pathway is an effective option for cancer therapy due to the presence of cell-suicide weapons in mitochondria. However, anti-apoptotic proteins that are over-expressed in the mitochondria of many malignant tumors, such as Bcl-2 protein, could allow the cancer cells to evade apoptosis, greatly reducing the efficacy of this type of chemotherapy. Here, we constructed a hierarchical targeted delivery system that can deliver siRNA and chemotherapeutic agents sequentially to tumor cells and mitochondria. In detail, the copolymer TPP-CP-LND (TCPL) was synthesized by the mitochondria-targeting ligand triphenylphosphine (TPP) and therapeutic drug lonidamine (LND) conjugated to the polyethyleneimine in chitosan-graft-PEI (CP), and then complexed with siRNA. Followed, the complexes were coated with poly(acrylic acid)-polyethylene glycol-folic acid (PPF) copolymer to form a hierarchical targeted co-delivery system (TCPL/siRNA/PPF NPs). The TCPL/siRNA/PPF NPs had a neutral surface charge, were stable in plasma and exhibited pH-responsive shell separation. Remarkably, the TCPL/siRNA/PPF NPs simultaneously released siBcl-2 into the cytoplasm and delivered LND to mitochondria in the same cancer cell after FA-directed internalization, and even synergistically activated mitochondria apoptosis pathway. This work demonstrated the potential of RNA-interference and mitochondria-targeted chemotherapeutics to collaboratively stimulate the mitochondria apoptosis pathway for cancer therapy.
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~1-1.5 mmol/g Capacity (Phosphor)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
6-Bromohexanoic acid, 97%
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%
Sigma-Aldrich
Rhodamine 123, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
DCC, 1.0 M in methylene chloride
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)