Merck
  • Two macrocyclic polyamines as modulators of metal-mediated Aβ40 aggregation.
Two macrocyclic polyamines as modulators of metal-mediated Aβ40 aggregation.
Integrative biology : quantitative biosciences from nano to macro (2015-05-15)
Yanfei Yang, Tingting Chen, Shajun Zhu, Xuefang Gu, Xueping Jia, Yapeng Lu, Li Zhu
ABSTRACT
Dysfunctional interactions of amyloid-β (Aβ) with Zn and Cu ions are proved to be related to the etiology of Alzheimer's disease (AD). Disruption of these metal-Aβ interactions using metal chelators holds considerable promise as a therapeutic strategy to combat this incurable disease. Herein, we report that two cyclam derivatives (L1 and L2) are capable of modulating Zn(2+)/Cu(2+)-mediated Aβ40 aggregation, reactive oxygen species (ROS) production, and neurotoxicity. These chelators were found to inhibit the metal-induced Aβ40 aggregation, dissociate metal-Aβ40 aggregates and restore the metal-induced β-sheet structure of Aβ40 to its random coil conformation, as observed by BCA protein assay, thioflavin T fluorescence and circular dichroism spectroscopy. Moreover, preliminary investigation of SH-SY5Y cells indicates that L1 and L2 can diminish the neurotoxicity of metal-Aβ40 species, control metal-Aβ40-triggered ROS production and protect cells against apoptosis. These observations warrant the further investigations of L1 and L2 as potential anti-AD agents.
Sigma-Aldrich
Copper(II) chloride, powder, 99%
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(II) chloride, 97%
Sigma-Aldrich
Thioflavin T, used as stain for amyloid