Merck
  • Organic anion transporter 4 (OAT 4) modifies placental transfer of perfluorinated alkyl acids PFOS and PFOA in human placental ex vivo perfusion system.
Organic anion transporter 4 (OAT 4) modifies placental transfer of perfluorinated alkyl acids PFOS and PFOA in human placental ex vivo perfusion system.
Placenta (2015-08-26)
M Kummu, E Sieppi, J Koponen, L Laatio, K Vähäkangas, H Kiviranta, A Rautio, P Myllynen
ABSTRACT
Perfluorinated alkyl acids (PFAAs) are widely used in industry and consumer products. Pregnant women are exposed to PFAAs and their presence in umbilical cord blood represents fetal exposure. Interestingly, PFAAs are substrates for organic anion transporters (OAT) of which OAT4 is expressed in human placenta. To evaluate the contribution of OAT4 and ATP-binding cassette transporter G2 (ABCG2) proteins in the transplacental transfer of perfluoro octane sulfonate (PFOS) and perfluoro octanoate (PFOA) an ex vivo dual recirculating human placental perfusion was used. Altogether 8 placentas from healthy mothers with uncomplicated pregnancies were successfully perfused. Both PFOS and PFOA crossed the placenta as suggested by in vivo data in the literature. The expression of OAT4 and ABCG2 proteins were studied by immunoblotting and correlation with the transfer index %(TI %) of PFOS and PFOA at 120 and 240 min (n = 4) was studied. The expression of OAT4 was in negative correlation with TI % of PFOA (R(2) = 0.92, p = 0.043) and PFOS (R(2) = 0.99, p = 0.007) at 120 min while at 240 min the correlation was statistically significant only with PFOA. The expression of ABCG2 did not correlate with TI% of PFOS or PFOA. Data obtained in this study suggest the involvement of OAT4 in placental passage of PFAAs. Placental passage of PFOS and PFOA is modified by the transporter protein OAT4 but not by ABCG2. This is the first study indicating that OAT4 may decrease the fetal exposure to PFAAs and protect the fetus after maternal exposure to PFAAs but further studies are needed to confirm our findings.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)