Merck
  • Effects of Sevoflurane on Young Male Adult C57BL/6 Mice Spatial Cognition.
Effects of Sevoflurane on Young Male Adult C57BL/6 Mice Spatial Cognition.
PloS one (2015-08-19)
Jianhui Liu, Xiaoqing Zhang, Wei Zhang, Guojun Gu, Peijun Wang
ABSTRACT
Inhalation anesthetics are reported to affect cognition in both animals and humans. The influence of inhalation anesthetics in learning and memory are contradictory. We therefore investigated the effects of sevoflurane anesthesia with different durations on cognitive performance and the levels of NMDA receptor subunit NR2B, phosphorylated ERK1/2 (p-ERK1/2) and activated caspase3 in mouse hippocampus. We anaesthetized eight-week old male C57BL/6 mice with 2.5% sevoflurane for durations ranging from one to four hours. Non-anaesthetized mice served as controls. Mice exposed to sevoflurane for one to three hours showed improved performance, whereas mice with exposure up to four hours displayed similar behavioral performance as control group. NR2B was increased both at 24h and at two weeks post sevoflurane exposure in all groups. The p-ERK1/2: total ERK1/2 ratio increased at 24h in all anesthesia groups. The ratio remained elevated at two weeks in groups with two- to four-hour exposure. Activated caspase3 was detected elevated at 24h in groups with two- to four-hour exposure. The elevated trend of activated caspase3 was still detectable at two weeks in groups with three- to four-hour exposure. At two weeks post anesthesia, the typical morphology associated with apoptotic cells was observed in the hippocampus of mice exposed to four hours of sevoflurane. Our results indicate that 2.5% sevoflurane exposure for one to three hours improved spatial cognitive performance in young adult mice. The cognitive improvement might be related to the increase of NR2B, the p-ERK1/2: total ERK1/2 ratio in hippocampus. However, exposure to sevoflurane for four hours caused neurotoxicity due to caspase3 activation and apoptosis.
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use