Merck
  • Identification and characterization of proteins with phenoloxidase-like activities in the sea urchin Strongylocentrotus nudus.
Identification and characterization of proteins with phenoloxidase-like activities in the sea urchin Strongylocentrotus nudus.
Fish & shellfish immunology (2015-09-01)
Yuhui Cheng, Jingwei Jiang, Ying Dong, Zunchun Zhou
ABSTRACT
Three proteins with PO-like activities in the coelomocytes of sea urchin Strongylocentrotus nudus were identified using electrophoretic method and named as SnPO1, SnPO2 and SnPO3 according to their molecular mass from high to low. The SnPOs were characterized for substrate specificity and the effects of temperature, pH, divalent metal ions and inhibitors on PO activities. They showed oxidative activities to L-3,4-dihydroxyphenylalanine. (l-DOPA), dopamine and hydroquinone, but failed to oxidize tyrosine, which illustrated the three proteins had laccase-like PO activities. The optimum temperature for the activities of SnPO1, SnPO2 and SnPO3 was 75 °C, 70 °C, 40 °C, and the optimum pH was 7.0, 9.0, 8.0, respectively. The SnPOs were notably activated after being incubated in boiled water for 60 min, suggesting that the three proteins are thermophilic. The activity of SnPO1 was greatly enhanced by Cu(2+), Mn(2+) and Fe(2+) and inhibited by Pb(2+), Cd(2+), EDTA, DETC, sodium sulfite and ascorbic acid, but SnPO2 and SnPO3 were not obviously affected by Pb(2+) and Cd(2+), suggesting the three proteins are copper-containing, and the catalytic properties of SnPO1 might be different from those of SnPO2 and SnPO3. Taken together, SnPO1, SnPO2 and SnPO3 might play different roles in the immune and physiological processes of S. nudus.
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)