Merck
  • Preoperative Detection and Intraoperative Visualization of Brain Tumors for More Precise Surgery: A New Dual-Modality MRI and NIR Nanoprobe.
Preoperative Detection and Intraoperative Visualization of Brain Tumors for More Precise Surgery: A New Dual-Modality MRI and NIR Nanoprobe.
Small (Weinheim an der Bergstrasse, Germany) (2015-06-11)
Chunyan Li, Limin Cao, Yejun Zhang, Peiwei Yi, Mao Wang, Bo Tan, Zongwu Deng, Dongmin Wu, Qiangbin Wang
ABSTRACT
In clinical practice, it is difficult to identify tumor margins during brain surgery due to its inherent infiltrative character. Herein, a unique dual-modality nanoprobe (Gd-DOTA-Ag2S QDs, referred as Gd-Ag2S nanoprobe) is reported, which integrates advantages of the deep tissue penetration of enhanced magnetic resonance (MR) imaging of Gd and the high signal-to-noise ratio and high spatiotemporal resolution of fluorescence imaging in the second near-infrared window (NIR-II) of Ag2S quantum dots (QDs). Due to the abundant tumor angiogenesis and the enhanced permeability and retention effect in the tumor, a brain tumor (U87MG) in a mouse model is clearly delineated in situ with the help of the Gd assisted T1 MR imaging and the intraoperative resection of the tumor is precisely accomplished under the guidance of NIR-II fluorescence imaging of Ag2S QDs after intravenous injection of Gd-Ag2S nanoprobe. Additionally, no histologic changes are observed in the main organs of the mouse after administration of Gd-Ag2S nanoprobe for 1 month, indicating the high biocompatibility of the nanoprobe. We expect that such a novel "Detection and Operation" strategy based on Gd-Ag2S nanoprobe is promising in future clinical applications.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
1-Decanethiol, 96%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
1-Decanethiol, 99%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Ethyl bromoacetate, reagent grade, 98%
Sigma-Aldrich
Gadolinium(III) chloride hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
1-Dodecanethiol, ≥98%
Sigma-Aldrich
tert-Butyl bromoacetate, 98%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Gadolinium(III) chloride hexahydrate, 99% (titration)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)