Merck
  • Physicochemical stability profile of Tulane virus: a human norovirus surrogate.
Physicochemical stability profile of Tulane virus: a human norovirus surrogate.
Journal of applied microbiology (2015-06-25)
S E Arthur, K E Gibson
ABSTRACT
Human norovirus (HuNoV) is estimated to cause 19-21 million illnesses each year in the US. A major limitation in HuNoV research is the lack of an in vitro culture system; therefore, surrogate viruses including murine norovirus (MNV) and feline calicivirus (FCV) are used to study HuNoV. Here, we aim to establish the physiochemical properties of Tulane virus (TV)—a newer HuNoV surrogate. For thermal inactivation, TV was exposed to 37°C for 2 h, and 56, 63 and 72°C for 30 min. For ethanol tolerance, TV was treated with 60, 70 and 90% ethanol at room temperature (RT) for 5 min. Tulane virus pH stability at pH 2, 3, 7, 9 and 10 was performed at RT for 90 min. At 37°C, there was no significant reduction in TV after 2 h. However, at 56, 63 and 72°C, D-values of 4·03, 1·18, and 0·24 min, were calculated respectively. The D-values obtained for TV ethanol tolerance were 1·46, 1·93, and 0·35 min at 60, 70 and 90% respectively. Less than 1 log10 plaque forming units (PFU) reduction was observed for TV at all pH levels except pH 10 where about a 2-log10 PFU reduction was observed. Tulane virus was also tolerant to chlorine disinfection on a solid surface with D-values of 15·82 and 5·42 min at 200 and 1000 ppm respectively. Tulane virus is likely a suitable surrogate to study HuNoV thermal stability as well as ethanol tolerance below 90%. Tulane virus also is a promising surrogate to study HuNoV pH stability and chlorine tolerance. Based on current work, in vitro studies demonstrate that TV is an overall more conservative and suitable surrogate for the study of HuNoV physicochemical properties.
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium Thiosulfate Solution, 2 g/dL in deionized water