Merck
  • Neurobehavioral Differences Between Mice Receiving Distinct Neuregulin Variants as Neonates; Impact on Sensitivity to MK-801.
Neurobehavioral Differences Between Mice Receiving Distinct Neuregulin Variants as Neonates; Impact on Sensitivity to MK-801.
Current molecular medicine (2015-03-31)
T Kato, Y Abe, S Hirokawa, Y Iwakura, M Mizuno, H Namba, H Nawa
ABSTRACT
Neuregulin-1 (NRG1) is a well-recognized risk gene for schizophrenia and is often implicated in the neurodevelopmental hypothesis of this illness. Alternative splicing and proteolytic processing of the NRG1 gene produce more than 30 structural variants; however, the neuropathological roles of individual variants remain to be characterized. On the basis of the neurodevelopmental hypothesis of schizophrenia, we administered eNRG1 (0.1~1.0 μg/g), a core epidermal growth factor-like (EGF) domain common for all splicing NRG1 variants, to neonatal mice and compared their behavioral performance with mice challenged with a full mature form of type 1 NRG1 variant. During the neonatal stage, recombinant eNRG1 protein administrated from the periphery passed the blood-brain barrier and activated its receptor (ErbB4) in the brain. In adults, the mice receiving the highest dose exhibited lower locomotor activity and deficits in prepulse inhibition and tonedependent fear learning, although the hearing reduction of the eNRG1-treated mice may explain these behavioral deficits. Neonatal eNRG1 treatment also significantly potentiated MK-801-driven locomotor activity in an eNRG1 dose-dependent manner. In parallel eNRG1 treatment enhanced MK-801-driven c-Fos induction and decreased immunoreactivity for NMDA receptor subunits in adult brain. In contrast, mice that had been treated with the same molar dose of a full mature form of type 1 NRG1 as neonates did not exhibit hypersensitivity to MK-801. However, both animal models exhibited similar hypersensitivity to methamphetamine. Collectively, our findings suggest that aberrant peripheral NRG1 signals during neurodevelopment alter later behavioral traits and auditory functions in the NRG1 subtype-dependent manner.
Sigma-Aldrich
EGF from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis