Merck
  • A new method and tool for detection and quantification of PM oxidative potential.
A new method and tool for detection and quantification of PM oxidative potential.
Environmental science and pollution research international (2015-04-24)
Francesco Ciriello, Maurizio Gualtieri, Eleonora Longhin, Riccardo Ruffo, Marina Camatini, Paolo Parenti
ABSTRACT
Airborne particulate matter (PM) contains several quinones, which are able to generate reactive oxygen species impacting on cell viability. A method able to detect and quantify PM oxidative potential, based on the cytochrome c (cyt-c) reduction by means of superoxide anion produced through quinones redox cycling in the presence of reducing agents, is here described. Tris(2-carboxyethyl)phosphine resulted to be the most efficient reducing agent among the ones tested. The procedure included rapid particles extraction, followed by two alternative analytical methods, a spectrophotometric assay based on the initial rate of cyt-c reduction at 550 nm, and an amperometric assay, based on self-assembled monolayers modified gold electrodes. The smallest amount of PM needed to obtain an evaluable signal is 2 μg. The described procedure may represent a starting point to develop devices for PM measurements in polluted atmospheric environments.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Phosphorus, red, ≥97.0%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Phosphorus, red, ≥99.99% trace metals basis
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
11-Mercaptoundecanoic acid, 95%
Sigma-Aldrich
11-Mercaptoundecanoic acid, 98%
Sigma-Aldrich
Anthraquinone, 97%
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride, flakes, 98%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
11-Mercapto-1-undecanol, 97%
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Manganese(II) chloride, powder and chunks, ≥99% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
11-Mercapto-1-undecanol, 99%
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O