Merck
  • Functionalized nanocarrier combined seizure-specific vector with P-glycoprotein modulation property for antiepileptic drug delivery.
Functionalized nanocarrier combined seizure-specific vector with P-glycoprotein modulation property for antiepileptic drug delivery.
Biomaterials (2015-10-09)
Jiansheng Liu, Yajing He, Jun Zhang, Jiajia Li, Xiangrong Yu, Zhonglian Cao, Fanmin Meng, Yuwu Zhao, Xunyi Wu, Teng Shen, Zhen Hong
ABSTRACT
Despite optimal therapeutic regimen with currently available antiepileptic drugs (AEDs), approximately a third of epilepsy patients remain drug refractory. Region-specific overexpression of multidrug efflux transporters at the blood-brain barrier, such as P-glycoprotein (P-gp), might contribute to multidrug resistance (MDR) by reducing target concentrations of AEDs. Therefore, development of nanomedicine that can modulate P-gp function as well as facilitate targeted AEDs delivery represents a promising strategy for epilepsy intervention. To achieve this, we sought to exploit the possibility of combination of active targeting function of tryptophan by transporter-mediated endocytosis and overcoming MDR by Pluronic block copolymers. Herein, a tryptophan derivate (TD) functionalized Pluronic P123/F127 mixed micelles encapsulating LTG (TD-PF/LTG) was developed to promote AEDs delivery to epileptogenic focus. TD-PF/LTG was about 20 nm in diameter with a spherical shape and high encapsulation efficiency. A rat epilepsy model with pilocarpine was established to evaluate the brain penetration efficiency of the LTG-incorporated polymeric micellar formulation, compared with free LTG formulations. Studies showed that TD-PF/LTG was more efficient than PF/LTG as well as free LTG in delivering the drug to the brain, especially the hippocampus. The enhanced targeted delivery could be ascribed to the increased tryptophan uptake at epileptogenic focus as well as P-gp modulation property of the nanomaterial. Taken together, TD-conjugated Pluronic micelles showed promising potential as a nanoplatform for the delivery of AEDs in refractory epilepsy.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
(Benzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Diethyl ether
SAFC
Glycine
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor