Merck
  • Comparison of the applicability of mass spectrometer ion sources using a polarity- molecular weight scattergram with a 600 sample in-house chemical library.
Comparison of the applicability of mass spectrometer ion sources using a polarity- molecular weight scattergram with a 600 sample in-house chemical library.
European journal of mass spectrometry (Chichester, England) (2015-07-17)
Natsuhiko Sugimura, Asami Furuya, Takahiro Yatsu, Toshimichi Shibue
ABSTRACT
To provide a practical guideline for the selection of a mass spectrometer ion source, we compared the applicability of three types of ion source: direct analysis in real time (DART), electrospray ionization (ESI) and fast atom bombardment (FAB), using an in-house high-resolution mass spectrometry sample library consisting of approximately 600 compounds. The great majority of the compounds (92%), whose molecular weights (MWs) were broadly distributed between 150 and 1000, were detected using all the ion sources. Nevertheless, some compounds were not detected using specific ion sources. The use of FAB resulted in the highest sample detection rate (>98%), whereas the detection rates obtained using DART and ESI were slightly lower (>96%). A scattergram constructed using MW and topological polar surface area (tPSA) as a substitute for molecular polarity showed that the performance of ESI was weak in the low-MW (<400), low-polarity (tPSA<60) area, whereas the performance of DART was weak in the high-MW (>800) area. These results might provide guidelines for the selection of ion sources for inexperienced mass spectrometry users.
Sigma-Aldrich
Carboxyamidotriazole, ≥98% (HPLC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
3-Nitrobenzyl alcohol, 98%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%