Merck
  • Chromatographic behavior of small organic compounds in low-temperature high-performance liquid chromatography using liquid carbon dioxide as the mobile phase.
Chromatographic behavior of small organic compounds in low-temperature high-performance liquid chromatography using liquid carbon dioxide as the mobile phase.
Journal of separation science (2015-04-29)
Tomohiro Motono, Takashi Nagai, Shinya Kitagawa, Hajime Ohtani
ABSTRACT
Low-temperature high-performance liquid chromatography, in which a loop injector, column, and detection cell were refrigerated at -35ºC, using liquid carbon dioxide as the mobile phase was developed. Small organic compounds (polyaromatic hydrocarbons, alkylbenzenes, and quinones) were separated by low-temperature high-performance liquid chromatography at temperatures from -35 to -5ºC. The combination of liquid carbon dioxide mobile phase with an octadecyl-silica (C18 ) column provided reversed phase mode separation, and a bare silica-gel column resulted in normal phase mode separation. In both the cases, nonlinear behavior at approximately -15ºC was found in the relationship between the temperature and the retention factors of the analytes (van't Hoff plots). In contrast to general trends in high-performance liquid chromatography, the decrease in temperature enhanced the separation efficiency of both the columns.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethylbenzene, anhydrous, 99.8%
Sigma-Aldrich
1,4-Naphthoquinone, 97%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Anthraquinone, 97%
Sigma-Aldrich
Propylbenzene, 98%
Sigma-Aldrich
Propylbenzene, ≥99.0% (GC)
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%