Merck
  • Pharmaceutical residues in water and sediment of Msunduzi River, KwaZulu-Natal, South Africa.
Pharmaceutical residues in water and sediment of Msunduzi River, KwaZulu-Natal, South Africa.
Chemosphere (2015-05-04)
Solomon Matongo, Grace Birungi, Brenda Moodley, Patrick Ndungu
ABSTRACT
The little data about pharmaceutical residue contamination in African water bodies motivated our study on the occurrence of pharmaceutical residues in the water and sediment of Msunduzi River in the KwaZulu-Natal province of South Africa; and in the Darvill wastewater treatment plant found in Msunduzi catchment. Samples collected along the River and wastewater treatment plant were extracted and analysed for pharmaceutical residues selected based on statistics of drug usage in South Africa i.e. antipyretics, antibiotics, caffeine, an antiepileptic and an antipsychotic drug were determined using HPLC-MS/MS. In all the matrices investigated, the antipyretic ibuprofen had the highest concentration of up to 117 μg L(-1), 84.60 μg L(-1) and 659 ng g(-1) in wastewater, surface water and sediment respectively. Antibiotics were detected in generally low concentrations of<10 μg L(-1) in surface water samples and up to 34.50 μg L(-1) in wastewater; moreover they were not completely removed during wastewater treatment. The percentage removal efficiency of the studied group was 6.55-98.00% for antipyretics, 73.33-98.90% for antibiotics, 48.80% for the anti-epileptic drug and 86.40% for Caffeine. Clozapine exhibited a negative removal. In surface water, Henley dam exhibited a high concentration of the pharmaceutical residues and the highest concentration of metronidazole in sediment (up to 1253.50 ng g(-1)) detected. Metronidazole was only detected in sediment and bio-solids.
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Ibuprofen
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture