Merck
  • A High-Throughput UHPLC-QqQ-MS Method for Polyphenol Profiling in Rosé Wines.
A High-Throughput UHPLC-QqQ-MS Method for Polyphenol Profiling in Rosé Wines.
Molecules (Basel, Switzerland) (2015-05-06)
Marine Lambert, Emmanuelle Meudec, Arnaud Verbaere, Gérard Mazerolles, Jérémie Wirth, Gilles Masson, Véronique Cheynier, Nicolas Sommerer
ABSTRACT
A rapid, sensitive and selective analysis method using Ultra High Performance Liquid Chromatography coupled to triple-quadrupole Mass Spectrometry (UHPLC-QqQ-MS) has been developed for the quantification of polyphenols in rosé wines. The compound detection being based on specific MS transitions in Multiple Reaction Monitoring (MRM) mode, the present method allows the selective quantification of up to 152 phenolic and two additional non-phenolic wine compounds in 30 min without sample purification or pre-concentration, even at low concentration levels. This method was repeatably applied to a set of 12 rosé wines and thus proved to be suitable for high-throughput and large-scale metabolomics studies.
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Protocatechuic acid ethyl ester, 97%
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
2-(4-Hydroxyphenyl)ethanol, 98%
Sigma-Aldrich
3-(2-Hydroxyethyl)indole, 97%
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)