Merck
  • In vivo and ex vivo proofs of concept that cetuximab conjugated vitamin E TPGS micelles increases efficacy of delivered docetaxel against triple negative breast cancer.
In vivo and ex vivo proofs of concept that cetuximab conjugated vitamin E TPGS micelles increases efficacy of delivered docetaxel against triple negative breast cancer.
Biomaterials (2015-06-18)
Rajaletchumy Veloo Kutty, Sing Ling Chia, Magdiel I Setyawati, Madaswamy S Muthu, Si-Shen Feng, David Tai Leong
ABSTRACT
In this study we examined the efficacy of our micellar system in xenograft models of triple negative breast cancers and explored the effect of the micelles on post-treatment tumours in order to elucidate the mechanism underlying the nanomedicine treatment in oncology. Here, we developed docetaxel-loaded vitamin E D-α-tocopheryl polyethylene glycol succinate (TPGS) micelles, of which the surface modified with cetuximab ligands for targeting epidermal growth factor receptors (EGFR) that are overexpressed in MDA-MB-231 breast cancer cells. The targeting micelles accumulated in the tumours immediately after the intravenous injection and retained for at least 24 h. The successful delivery of docetaxel into the tumours by the targeting micelles was shown by the greater degree of tumour growth inhibition than that for Taxotere(®) after the 15-day treatment. Furthermore, the explanted tumour culture study involving gene analysis and immunohistochemistry staining indicated that the in vivo micelle treatment induced cell cycle arrest and attenuated cell proliferation. In addition, the targeting and non-targeting micellar formulations brought about anti-angiogenesis and anti-migration effects. Overall, both the in vivo and ex vivo data increased the confidence that our micellar formulations effectively targeted and inhibited EGFR-overexpressing MDA-MB-231 tumours.
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
D-Luciferin, synthetic, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-Luciferin, synthetic
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
Eosin Y disodium salt, certified by the BSC
Sigma-Aldrich
Eosin Y disodium salt, ≥85 % by HPLC
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g