Merck
  • Diversity of black Aspergilli isolated from raisins in Argentina: Polyphasic approach to species identification and development of SCAR markers for Aspergillus ibericus.
Diversity of black Aspergilli isolated from raisins in Argentina: Polyphasic approach to species identification and development of SCAR markers for Aspergillus ibericus.
International journal of food microbiology (2015-06-27)
G Giaj Merlera, S Muñoz, I Coelho, L R Cavaglieri, A M Torres, M M Reynoso
ABSTRACT
Aspergillus section Nigri is a heterogeneous fungal group including some ochratoxin A producer species that usually contaminate raisins. The section contains the Series Carbonaria which includes the toxigenic species Aspergillus carbonarius and nontoxigenic Aspergillus ibericus that are phenotypically undistinguishable. The aim of this study was to examine the diversity of black aspergilli isolated from raisins and to develop a specific genetic marker to distinguish A. ibericus from A. carbonarius. The species most frequently found in raisins in this study were Aspergillus tubingensis (35.4%) and A. carbonarius (32.3%), followed by Aspergillus luchuensis (10.7%), Aspergillus japonicus (7.7%), Aspergillus niger (6.2%), Aspergillus welwitschiae (4.6%) and A. ibericus (3.1%). Based on inter-simple sequence repeat (ISSR) fingerprinting profiles of major Aspergillus section Nigri members, a sequence-characterized amplified region (SCAR) marker was identified. Primers were designed based on the conserved regions of the SCAR marker and were utilized in a PCR for simultaneous identification of A. carbonarius and A. ibericus. The detection level of the SCAR-PCR was found to be 0.01 ng of purified DNA. The present SCAR-PCR is rapid and less cumbersome than conventional identification techniques and could be a supplementary strategy and a reliable tool for high-throughput sample analysis.
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)