Merck
Role of matrix metalloproteinases in non-healing venous ulcers.
International wound journal (2013-10-30)
Bruno Amato, Guido Coretti, Rita Compagna, Maurizio Amato, Gianluca Buffone, Diego Gigliotti, Raffaele Grande, Raffaele Serra, Stefano de Franciscis
ABSTRACT
Chronic venous ulceration (CVU) of the lower limbs is a common condition affecting 1% of the adult population in Western countries, which is burdened with a high complication rate and a marked reduction in the quality of life often due to prolonged healing time. Several metalloproteinases (MMPs) such as MMP-9 together with neutrophil gelatinase-associated lipocalin (NGAL) appear to be involved in the onset and healing phases of venous ulcer, but it is still unclear how many biochemical components are responsible for prolonged healing time in those ulcers. In this study, we evaluate the role of MMP-1 and MMP-8 in long lasting and refractory venous ulcers. In a 2-year period we enroled 45 patients (28 female and 17 male, median age 65) with CVU. The enroled population was divided into two groups: group I were patients with non-healing ulcers (ulcers that had failed to heal for more than 2 months despite appropriate treatments) and group II were patients with healing ulcers (ulcers in healing phases). MMP-1 and MMP-8 were measured in fluids and tissues of healing and non-healing ulcers by means of enzyme-linked immunosorbent assay (ELISA) and Western blot analysis, respectively. In particular the patterns of the collagenases MMP-1 and MMP-8 in healing wounds were distinct, with MMP-8 appearing in significantly greater amounts especially in the non-healing group. Our findings suggest that MMP-1, and MMP-8 are overexpressed in long lasting CVU. Therefore, this dysregulation may represent the main cause of the pathogenesis of non-healing CVU.
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
BICINE, ≥99% (titration)
Sigma-Aldrich
BICINE, BioXtra, ≥99% (titration)