Merck
  • High Cytotoxicity and Apoptotic Effects of Natural Bioactive Benzofuran Derivative on the MCF-7 Breast Cancer Cell Line.
High Cytotoxicity and Apoptotic Effects of Natural Bioactive Benzofuran Derivative on the MCF-7 Breast Cancer Cell Line.
Combinatorial chemistry & high throughput screening (2015-05-01)
Afsane Soleimani, Jahanbakhsh Asadi, Faramarz Rostami-Charati, Roghaye Gharaei
ABSTRACT
This study was focused on evaluation of the cytotoxicity and apoptotic affects of benzofuran derivative on MCF-7 breast cancer cell line. This effective compound was isolated from the root of Petasites hybridus plant. For experiments, the MCF-7 cells were treated with several concentrations (0-500μM) of 1-(6-hydroxy-2- isopropenyl-1-benzofuran-5-yl)-1-ethanone 1 at different times. In this study, test of neutral red was also employed for cytotoxicity assay and quantity of P53, P21. Bax genes expression was analyzed using Real-Time PCR and ELISA techniques. Results show that compound 1 has cytotoxicity and apoptotic effects on Human breast cancer (Michigan Cancer Foundation-7) MCF-7 cells.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma