Merck
  • A cytoplasmic helix is required for pentamer formation of the Escherichia coli MscL mechanosensitive channel.
A cytoplasmic helix is required for pentamer formation of the Escherichia coli MscL mechanosensitive channel.
Journal of biochemistry (2015-02-24)
Chie Ando, Naili Liu, Kenjiro Yoshimura
ABSTRACT
Many membrane proteins such as ion channels are oligomers, but the determinants of the degree of oligomerization are not fully understood. Mechanosensitive channel with large conductance (MscL), which is ubiquitous in bacteria, is a homopentamer with two transmembrane helices and a cytoplasmic helix in each subunit. The carboxyl-terminal cytoplasmic helices assemble into a pentameric bundle that resembles cartilage oligomeric matrix protein. To address the role of cytoplasmic helices in the pentamer formation of Escherichia coli MscL, we generated MscL constructs with various deletions at the carboxyl terminus and translated them in a cell-free system. Deletions of Leu-129 and the downstream sequence resulted in formation of various oligomers without preference to pentamers, suggesting that nearly the whole cytoplasmic helix is required for MscL pentamer formation.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium phosphate, 96%