Merck
  • Preparation and evaluation of biocompatible long-term radiopaque microspheres based on polyvinyl alcohol and lipiodol for embolization.
Preparation and evaluation of biocompatible long-term radiopaque microspheres based on polyvinyl alcohol and lipiodol for embolization.
Journal of biomaterials applications (2015-03-15)
Wen-Jing Meng, Xiao-Jing Lu, Huan Wang, Tian-Yuan Fan, Dai-Chao Cui, Shui-Sheng Zhang, Zhuo-Zhao Zheng, Hai-Tao Guan, Li Song, Ying-Hua Zou
ABSTRACT
The aim of this work was to develop long-term radiopaque microspheres (LRMs) by entrapping lipiodol in biocompatible polyvinyl alcohol with multiple emulsions chemical crosslinking method. The high content of lipiodol (0.366 g/mL) was hardly released from LRMs in vitro and the radiopacity could maintain at least 3 months after subcutaneous injection in mice without weakening. A series of tests was performed to evaluate the feasibility of LRMs for embolization. LRMs were proved to be smooth, spherical, and well dispersed with diameter range of 100-1200 μm. Young's modulus of LRMs was 55.39 ± 9.10 kPa and LRMs could be easily delivered through catheter without aggregating or clogging. No toxicity of LRMs was found to mouse L929 fibroblasts cells and only moderate inflammatory in surrounding tissue of mice was found after subcutaneous injection of LRMs. After LRMs were embolized in renal artery of a rabbit, the distribution and radiopacity of LRMs in vivo were easily detectable by X-ray fluoroscopy and computed tomography (CT) imaging, respectively. More accurate distribution of LRMs in embolized kidney and vessels could be detected by high-revolution visualization of micro-CT ex vivo. In conclusion, the LRMs were proved to be biocompatible and provide long-term radiopacity with good physical and mechanical properties for embolization.
Sigma-Aldrich
Dimethyl sulfide, natural, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfide, anhydrous, ≥99.0%
Sigma-Aldrich
Dimethyl sulfide, ≥99%
Sigma-Aldrich
Dimethyl sulfide, ≥99%, FCC, FG