Merck
  • Antimicrobial Electrospun Biopolymer Nanofiber Mats Functionalized with Graphene Oxide-Silver Nanocomposites.
Antimicrobial Electrospun Biopolymer Nanofiber Mats Functionalized with Graphene Oxide-Silver Nanocomposites.
ACS applied materials & interfaces (2015-05-20)
Andreia F de Faria, François Perreault, Evyatar Shaulsky, Laura H Arias Chavez, Menachem Elimelech
ABSTRACT
Functionalization of electrospun mats with antimicrobial nanomaterials is an attractive strategy to develop polymer coating materials to prevent bacterial colonization on surfaces. In this study we demonstrated a feasible approach to produce antimicrobial electrospun mats through a postfabrication binding of graphene-based nanocomposites to the nanofibers' surface. A mixture of poly(lactide-co-glycolide) (PLGA) and chitosan was electrospun to yield cylindrical and narrow-diameter (356 nm) polymeric fibers. To achieve a robust antimicrobial property, the PLGA-chitosan mats were functionalized with graphene oxide decorated with silver nanoparticles (GO-Ag) via a chemical reaction between the carboxyl groups of graphene and the primary amine functional groups on the PLGA-chitosan fibers using 3-(dimethylamino)propyl-N'-ethylcarbodiimide hydrochloride and N-hydroxysuccinimide as cross-linking agents. The attachment of GO-Ag sheets to the surface of PLGA-chitosan fibers was successfully revealed by scanning and transmission electron images. Upon direct contact with bacterial cells, the PLGA-chitosan mats functionalized with GO-Ag nanocomposites were able to effectively inactivate both Gram-negative (Escherichia coli and Pseudomonas aeruginosa) and Gram-positive (Staphylococcus aureus) bacteria. Our results suggest that covalent binding of GO-Ag nanocomposites to the surface of PLGA-chitosan mats opens up new opportunities for the production of cost-effective, scalable, and biodegradable coating materials with the ability to hinder microbial proliferation on solid surfaces.
Sigma-Aldrich
Sodium phosphate dibasic, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use