Merck
  • Comparison of accelerated solvent extraction and quick, easy, cheap, effective, rugged and safe method for extraction and determination of pharmaceuticals in vegetables.
Comparison of accelerated solvent extraction and quick, easy, cheap, effective, rugged and safe method for extraction and determination of pharmaceuticals in vegetables.
Journal of chromatography. A (2015-06-13)
Ya-Hui Chuang, Yingjie Zhang, Wei Zhang, Stephen A Boyd, Hui Li
ABSTRACT
Land application of biosolids and irrigation with reclaimed water in agricultural production could result in accumulation of pharmaceuticals in vegetable produce. To better assess the potential human health impact from long-term consumption of pharmaceutical-contaminated vegetables, it is important to accurately quantify the amount of pharmaceuticals accumulated in vegetables. In this study, a quick, easy, cheap, effective, rugged and safe (QuEChERS) method was developed and optimized to extract multiple classes of pharmaceuticals from vegetables, which were subsequently quantified by liquid chromatography coupled to tandem mass spectrometry. For the eleven target pharmaceuticals in celery and lettuce, the extraction recovery of the QuEChERS method ranged from 70.1 to 118.6% with relative standard deviation <20%, and the method detection limit was achieved at the levels of nanograms of pharmaceuticals per gram of vegetables. The results revealed that the performance of the QuEChERS method was comparable to, or better than that of accelerated solvent extraction (ASE) method for extraction of pharmaceuticals from plants. The two optimized extraction methods were applied to quantify the uptake of pharmaceuticals by celery and lettuce growing hydroponically. The results showed that all the eleven target pharmaceuticals could be absorbed by the vegetables from water. Compared to the ASE method, the QuEChERS method offers the advantages of short time and reduced costs of sample preparation, and less amount of organic solvents used. The established QuEChERS method could be used to determine the accumulation of multiple classes of pharmaceutical residues in vegetables and other plants, which is needed to evaluate the quality and safety of agricultural produce consumed by humans.
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®